Mobile stand, suitable for stirring drives MAXdrive and FABdrive as well as control units MAXcontrol and FABcontrol, large surface f or vessel and containers up to 57 cm diameter, load up to 250 kg, robust design made from resistant stainless steel, 1 * 1 items
Catalog # 442-0819
Supplier: 2MAG
Frequently Bought Together