ContactUsLinkComponent
Magnetic stirrer, wear-free with 1 stirring position, for up to 1,000 ml stirring volume.1 stirring position, 100% maintenance-free and wear-free by inductive 2mag-Magnetic- 1 * 1 items
Catalog # TMAG40300_CH
Supplier:  2MAG
Magnetic stirrer, wear-free with 1 stirring position, for up to 1,000 ml stirring volume.1 stirring position, 100% maintenance-free and wear-free by inductive 2mag-Magnetic- 1 * 1 items
Catalog # TMAG40300_CH
Supplier:  2MAG
Frequently Bought Together