Packaging and Storage Boxes
Transparent, 44×22×18 mm, PS
SEMA1984
: SEMADENI
These boxes made of polystyrene (PS) are crystal clear with a good closing tight fitting lid.
- Clear for easy product identification
- The lids are either mounted or not mounted
- Stackable
- Colour:Transparent
- Material:PS
- Seq:26
- W×D×H:44×22×18 mm