Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 16, excitation BP 485/20, beam splitter FT 510, emission LP 515
Catalog # ZEIS4880169901000
Supplier:   ZEISS
Filter sets
Catalog # ZEIS4880169901000
Supplier:  ZEISS
Filter sets
Filter set 16, excitation BP 485/20, beam splitter FT 510, emission LP 515
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Filter set 16, excitation BP 485/20, beam splitter FT 510, emission LP 515
  • Seq:
    04