ContactUsLinkComponent
Tubing, Dual Layer PEEK, 1/16'' x 0.25 mm ID, solid blue outside, natural inside, 10 m/pkg 1 * 10 m
Catalog # JOURJR-TD-6009-M10
Supplier:  VICI JOUR RESEARCH
Tubing, Dual Layer PEEK, 1/16'' x 0.25 mm ID, solid blue outside, natural inside, 10 m/pkg 1 * 10 m
Catalog # JOURJR-TD-6009-M10
Supplier:  VICI JOUR RESEARCH
Frequently Bought Together