Orders LinkContactUsLinkComponent
Cable ties, Blitzbinder®
Cable ties, Blitzbinder®
Yellow, 120 mm
Catalog # SCAC6200-GELB
Cable ties, Blitzbinder®
Catalog # SCAC6200-GELB
Cable ties, Blitzbinder®
Cable ties, Blitzbinder®
Yellow, 120 mm
Price
103,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
PE cable ties for quick and safe closure of bags, sacks etc.
Available in different sizes and colours.
Specifications
  • Colour:
    Yellow
  • Dimensions:
    120 mm
  • Seq:
    03