Orders LinkContactUsLinkComponent
Embedding cassettes
Embedding cassettes
Embedding cassettes, White
Catalog # MARI5253104
Supplier:   MARIENFELD
Embedding cassettes
Catalog # MARI5253104
Supplier:  MARIENFELD
Embedding cassettes
Embedding cassettes
Embedding cassettes, White
Price
82,89 €
Log in for availability
1 * 500 items
Product Details & Documents
Plastic (POM).
  • Resistant against the solvents usually used in laboratories
  • Different colours offer the possibility of colour coding (for example for different users, priorities)
  • One 45° inclined and two vertical marking areas
  • Roughness of the marking areas optimized to allow labelling by different systems
Specifications
  • Colour:
    White
  • Description:
    Embedding cassettes
  • Seq:
    01