Packaging and Storage Boxes
Transparent, 155×138×59 mm, PS
SEMA1738
: SEMADENI
These boxes made of polystyrene (PS) are crystal clear with a good closing tight fitting lid.
- Clear for easy product identification
- The lids are either mounted or not mounted
- Stackable
- Colour:Transparent
- Material:PS
- Seq:18
- W×D×H:155×138×59 mm