Pipette tips
With collar, 100 - 1000 µl, -, Blue, Eppendorf, Nichiryo, Biohit, Brand, Gilson, Treff, Menarini, Bulk (10×1000)
: DELTALAB
Documents
- Packed:Bulk (10×1000)
- Sterile:-
- Volume:100 - 1000 µl
- Seq:15
- Tip style:With collar
- Tip colour:Blue
- For use with:Eppendorf, Nichiryo, Biohit, Brand, Gilson, Treff, Menarini