ContactUsLinkComponent
Elmasonic X-tra basic 1200 - ultrasonic cleaner Int.Dem. of tank (mm) 600x600x350, Volume: 126 liters, Serv. Cap. 97 1 * 1 items
  ELMA1027047
Elmasonic X-tra basic 1200 - ultrasonic cleaner Int.Dem. of tank (mm) 600x600x350, Volume: 126 liters, Serv. Cap. 97 1 * 1 items
  ELMA1027047
Frequently Bought Together