Orders LinkContactUsLinkComponent
Sample cups for analysers
Sample cups for auto sampler, PS, recommended for use with most aqueous solutions, 1,5 - 2 ml, Clear
  PERCB0119079
Sample cups for analysers
  PERCB0119079
Sample cups for analysers
Sample cups for auto sampler, PS, recommended for use with most aqueous solutions, 1,5 - 2 ml, Clear
245,00 €
1 * 1.000 items
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.
  • Capacity:
    1,5 - 2 ml
  • Colour:
    Clear
  • Description:
    Sample cups for auto sampler, PS, recommended for use with most aqueous solutions