Orders LinkContactUsLinkComponent
Sample cups for analysers
Sample cups for auto sampler, PTFE, recommended for use with most concentrated acids including HF and for analyses where the lowest detection levels are required, 2 ml, Clear
Catalog # PERCB0087600
Supplier:   PerkinElmer
Sample cups for analysers
Catalog # PERCB0087600
Supplier:  PerkinElmer
Sample cups for analysers
Sample cups for auto sampler, PTFE, recommended for use with most concentrated acids including HF and for analyses where the lowest detection levels are required, 2 ml, Clear
Price
792,00 €
Log in for availability
1 * 30 items
Product Details & Documents
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.
Specifications
  • Capacity:
    2 ml
  • Colour:
    Clear
  • Description:
    Sample cups for auto sampler, PTFE, recommended for use with most concentrated acids including HF and for analyses where the lowest detection levels are required