Orders LinkContactUsLinkComponent
Sample cups for analysers
Sample cups for analysers
Sample cups for auto sampler, PP, no known solvent at room temperature, 2,5 ml, Translucent
Catalog # PERCB3001566
Supplier:   PerkinElmer
Sample cups for analysers
Catalog # PERCB3001566
Supplier:  PerkinElmer
Sample cups for analysers
Sample cups for auto sampler, PP, no known solvent at room temperature, 2,5 ml, Translucent
Price
168,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Disposable.
These sample cups are of heavy-duty construction to prevent cracking or tipping. Their conical design allows for use of nearly the entire sample. For concentrated acids including HF for analyses.
Specifications
  • Capacity:
    2,5 ml
  • Colour:
    Translucent
  • Description:
    Sample cups for auto sampler, PP, no known solvent at room temperature