Orders LinkContactUsLinkComponent
Objectives
A-Planachromatic, Ph1, M27, N.A. of 0,25, WD of 4,5 mm
  ZEIS4210419910000
 :   ZEISS
Objectives
  ZEIS4210419910000
 :  ZEISS
Objectives
Objectives
A-Planachromatic, Ph1, M27, N.A. of 0,25, WD of 4,5 mm
1 * 1 items
  • Magnification:
    10×
  • Description:
    A-Planachromatic, Ph1, M27, N.A. of 0,25, WD of 4,5 mm
  • Seq:
    06