ContactUsLinkComponent
RTS 500 Adapter Stainless steel frame for holding up to six RTS 500 CECF devices for use with the Eppendorf MTP Thermoblock or RTS P roteoMaster Instrument. 1 * 1 items
Catalog # BTRABR1401901
Supplier:  biotechrabbit
RTS 500 Adapter Stainless steel frame for holding up to six RTS 500 CECF devices for use with the Eppendorf MTP Thermoblock or RTS P roteoMaster Instrument. 1 * 1 items
Catalog # BTRABR1401901
Supplier:  biotechrabbit
Frequently Bought Together