ContactUsLinkComponent
The microprocessor controlled ramp regulator PR 5 SR is a complex time/temperature control for the precision hotplates. Pressing the START key enables the programming of a set of a maximum of 5 ramps (each ramp consists of the ramp-time, constant tem 1 * 1 items
Catalog # GESTPR5SR
Supplier:  GESTIGKEIT HARRY
The microprocessor controlled ramp regulator PR 5 SR is a complex time/temperature control for the precision hotplates. Pressing the START key enables the programming of a set of a maximum of 5 ramps (each ramp consists of the ramp-time, constant tem 1 * 1 items
Catalog # GESTPR5SR
Supplier:  GESTIGKEIT HARRY
Frequently Bought Together