Orders LinkContactUsLinkComponent
Spatulas
Spatulas
180 mm, 20 mm, White
Catalog # COWI043.180
Supplier:   COWIE
Spatulas
Catalog # COWI043.180
Supplier:  COWIE
Spatulas
Spatulas
180 mm, 20 mm, White
Price
20,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Double-ended spatula/scraper combination made of PTFE, with fine chamfered ends.
Documents
Specifications
  • Colour:
    White
  • Length:
    180 mm
  • Blade width:
    20 mm