Orders LinkContactUsLinkComponent
Spatulas
Spatulas
120 mm, 12 mm, White
Catalog # COWI043.120
Supplier:   COWIE
Spatulas
Catalog # COWI043.120
Supplier:  COWIE
Spatulas
Spatulas
120 mm, 12 mm, White
Price
14,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Double-ended spatula/scraper combination made of PTFE, with fine chamfered ends.
Documents
Specifications
  • Colour:
    White
  • Length:
    120 mm
  • Blade width:
    12 mm