Kit conjugation AP-Antibody all-in-one. Used to conjugate and purify your antibody labels. Contains purification columns. To perform 2 conjugations of 100µg of antibody each. 1 * 1 KIT
Catalog # SOLUA-9105-001
Supplier: TriLink BioTechnologies
Frequently Bought Together