ContactUsLinkComponent
GE100, Standard-pH-elektrode for GPRT1400AN pH meter Measuring range: 0-14 pH, 0-80 C, not pressure resistant Electrolyte: 3mol/l KCL, refillable Miscellaneous: 1m cable, with cinch plug Scope of supply: pH-electrode, operating instructions Applicat 1 * 1 items
Catalog # GRSG102080
Supplier:  Greisinger
GE100, Standard-pH-elektrode for GPRT1400AN pH meter Measuring range: 0-14 pH, 0-80 C, not pressure resistant Electrolyte: 3mol/l KCL, refillable Miscellaneous: 1m cable, with cinch plug Scope of supply: pH-electrode, operating instructions Applicat 1 * 1 items
Catalog # GRSG102080
Supplier:  Greisinger
Frequently Bought Together