Orders LinkContactUsLinkComponent
Compact automatic burettes, Schellbach, BLAUBRAND®
25 ml, 0,05 ml, 785 mm, 0,03 ± ml
  612-7171
 :   Brand
Compact automatic burettes, Schellbach, BLAUBRAND®
  612-7171
 :  Brand
Compact automatic burettes, Schellbach, BLAUBRAND®
Compact automatic burettes, Schellbach, BLAUBRAND®
25 ml, 0,05 ml, 785 mm, 0,03 ± ml
140,00 €
1 * 1 items
Borosilicate glass 3.3 bottle, class AS, DE-M marking.
  • Calibrated to deliver 'TD, Ex'
  • Schellbach stripe
  • Detachable stopcock
 : DIN EN ISO 385
 : Please order burette clamps, supports and rubber bellows separately.
 : Supplied as burette tube, stopcock with precision tip, filling tube (PVC) and 1000 ml PE bottle
  • Capacity:
    25 ml
  • Length:
    785 mm
  • Division:
    0,05 ml
  • Tolerance:
    0,03 ± ml
Accessories
  •  
    Description
  • BRND23400
    Single bulb rubber bellows, orange coloured, with valve and tubing
    1 * 1 items - 16,10 €
  •  
    Description
    Capacity
  • BRND23728
    Bases for bottles
    1000 ml
    1 * 1 items - 7,80 €
  •  
    Description
  • BRND11525
    Spare filling tube for all compact automatic burettes, PVC
    1 * 1 items - 4,40 €
  •  
    Description
    Capacity
  • BRND23725
    Bases for bottles
    500 ml
    1 * 1 items - 6,60 €
  •  
    Description
    Length
  • BRND23875
    Pump head, clear, NS 29/32, fitting length 185 mm, nozzle outer Ø 7 mm, Boro 3.3
    185 mm
    1 * 1 items - 41,00 €
  •  
    Description
  • BRND11510
    Spare burette tip, clear borosilicate 3.3 glass, for 10 ml compact burettes and compact automatic burettes
    1 * 1 items - 6,30 €
  •  
    Description
  • BRND11805
    Spare burette stopcock, PTFE
    1 * 1 items - 46,00 €
  •  
    Description
  • BRND11512
    Spare burette tip, clear borosilicate 3.3 glass, for 50 ml compact burettes and compact automatic burettes
    1 * 1 items - 6,30 €
  •  
    Description
    Capacity
    Division
  • BRND23838
    Spare burette tubes, AR-Glas®
    50 ml
    0,1 ml
    1 * 1 items - 50,40 €
  •  
    Description
  • BRND11511
    Spare burette tip, clear borosilicate 3.3 glass, for 25 ml compact burettes and compact automatic burettes
    1 * 1 items - 6,30 €
  •  
    Description
    Capacity
    Division
  • BRND23833
    Spare burette tubes, AR-Glas®
    10 ml
    0,05 ml
    1 * 1 items - 50,40 €
  •  
    Description
    Capacity
    Division
  • BRND23836
    Spare burette tubes, AR-Glas®
    25 ml
    0,1 ml
    1 * 1 items - 50,40 €
  •  
    Description
    Capacity
  • BRND129055
    Reservoir bottle, LDPE, GL 25
    500 ml
    1 * 1 items - 4,10 €
  •  
    Description
    Capacity
  • BRND129060
    Reservoir bottle, LDPE, GL 28
    1000 ml
    1 * 1 items - 7,30 €