Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
10 ml, 10/19, 90 mm
Catalog # HIRS7560160
Supplier:   HIRSCHMANN
Volumetric flasks, class B
Catalog # HIRS7560160
Supplier:  HIRSCHMANN
Volumetric flasks, class B
Volumetric flasks, class B
10 ml, 10/19, 90 mm
Price
26,30 €
Log in for availability
1 * 2 items
Product Details & Documents
PMP, translucent, with PP NS-stopper.
Specifications
  • NS:
    10/19
  • Capacity:
    10 ml
  • Height:
    90 mm