Micro haematocrit capillaries, disposable
With sodium-heparin, 9 µl, 32 mm, - mm, - mm
Catalog # 720-2948
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Disposable capillaries for centrifugation of blood tests.
- Even and heavy wall thickness
- With printed colour-code
According to DIN 12846
: According to DIN 12846
: 100 pieces packed in a glass vial, 10 vials in a carton.
Specifications
- Ext.Ø:- mm
- Ø int.:- mm
- Description:With sodium-heparin, 9 µl
- Length:32 mm
- Seq:03