Orders LinkContactUsLinkComponent
Micro haematocrit capillaries, disposable
Micro haematocrit capillaries, disposable
With sodium-heparin, 9 µl, 32 mm, - mm, - mm
Catalog # 720-2948
Supplier:   HIRSCHMANN
Micro haematocrit capillaries, disposable
Catalog # 720-2948
Supplier:  HIRSCHMANN
Micro haematocrit capillaries, disposable
Micro haematocrit capillaries, disposable
With sodium-heparin, 9 µl, 32 mm, - mm, - mm
Price
150,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Disposable capillaries for centrifugation of blood tests.
  • Even and heavy wall thickness
  • With printed colour-code
According to DIN 12846
 : According to DIN 12846
 : 100 pieces packed in a glass vial, 10 vials in a carton.
Specifications
  • Ext.Ø:
    - mm
  • Ø int.:
    - mm
  • Description:
    With sodium-heparin, 9 µl
  • Length:
    32 mm
  • Seq:
    03