ScreenMates 1.4mL round-bottom storage tube, sterile, printed with alphanumeric (A1-H12) labels. Polypropylene. Racked in EcoTube re fill packs. 10 racks of 96 tubes. 1 * 960 items
Catalog # MATR4254
Supplier: THERMO MATRIX TECHNOLOGIES
Frequently Bought Together