Caps, ERLENMEYER-CAP
Silver, 37-39 mm
LUDI184115391
: LUDI
Aluminium, anodised.
- Springs made of chromium-nickel steel
- Springs clamp against the outer wall of the vessel
- Autoclavable
- For:37-39 mm
- Colour:Silver
- Seq:01