Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Filter set 20, excitation BP 546/12, beam splitter FT 560, emission BP 575-640
Catalog # ZEIS4880209901000
Supplier:   ZEISS
Filter sets
Catalog # ZEIS4880209901000
Supplier:  ZEISS
Filter sets
Filter set 20, excitation BP 546/12, beam splitter FT 560, emission BP 575-640
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Filter set 20, excitation BP 546/12, beam splitter FT 560, emission BP 575-640
  • Seq:
    05