Staining tray with lid, APET, 100 x 143 x 25 mm, 20 trays/case, Staining trays are available in either polypropylene or APET material. Polypropylene trays are microwavable for heat applications. The APET material has an optically clear bottom and is 1 * 20 items
Catalog # BITX731-005
Supplier: BIOTIX
Frequently Bought Together