Orders LinkContactUsLinkComponent
Objectives
Objectives
EC-Planachromatic Neofluar, Ph2, M27, N.A. of 0,5, WD of 2,0 mm
Catalog # ZEIS4203519910000
Supplier:   ZEISS
Objectives
Catalog # ZEIS4203519910000
Supplier:  ZEISS
Objectives
Objectives
EC-Planachromatic Neofluar, Ph2, M27, N.A. of 0,5, WD of 2,0 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    20×
  • Description:
    EC-Planachromatic Neofluar, Ph2, M27, N.A. of 0,5, WD of 2,0 mm
  • Seq:
    20