Cryoboxes
-, -, White, 75×75×50 mm
Catalog # RATI5160035
Supplier: RATIOLAB
Jump to:
Product Details & Documents
Cryoboxes, cardboard, for storage of cryovials at low temperatures.
- Waterproof coating
- Dividers must to be ordered separately
- Available in different heights and five colours
Specifications
- Compartments:-
- For:-
- W×D×H ext.:75×75×50 mm
- Colour:White