Filter sets
Filter set 38 (high efficiency), excitation BP 470/40 (HE), beam splitter FT 495 (HE), emission BP 525/50 (HE)
ZEIS4890389901000
: ZEISS
- Description:Filter set 38 (high efficiency), excitation BP 470/40 (HE), beam splitter FT 495 (HE), emission BP 525/50 (HE)
- Seq:09