Orders LinkContactUsLinkComponent
Objectives
EC-Planachromatic Neofluar, Ph2, M27, N.A. of 0,75, WD of 0,71 mm
  ZEIS4203619910000
 :   ZEISS
Objectives
  ZEIS4203619910000
 :  ZEISS
Objectives
Objectives
EC-Planachromatic Neofluar, Ph2, M27, N.A. of 0,75, WD of 0,71 mm
1 * 1 items
  • Magnification:
    40×
  • Description:
    EC-Planachromatic Neofluar, Ph2, M27, N.A. of 0,75, WD of 0,71 mm
  • Seq:
    22