Orders LinkContactUsLinkComponent
Objectives
Objectives
EC-Planachromatic Neofluar, Ph1, M27, N.A. of 0,3, WD of 5,5 mm
Catalog # ZEIS4203419911000
Supplier:   ZEISS
Objectives
Catalog # ZEIS4203419911000
Supplier:  ZEISS
Objectives
Objectives
EC-Planachromatic Neofluar, Ph1, M27, N.A. of 0,3, WD of 5,5 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    10×
  • Description:
    EC-Planachromatic Neofluar, Ph1, M27, N.A. of 0,3, WD of 5,5 mm
  • Seq:
    18