Cased IP66 Dispenser peristaltic pumps, programmable for batch dosing with calibration, speed ramp and run-back features, variable s peed/flow rate up to 1310 ml/min. and indication on digital display of speed, flow rate and tube size. Pumps can be p 1 * 1 items
Catalog # 181-0183
Supplier: VWR Collection
Frequently Bought Together