Orders LinkContactUsLinkComponent
Microscope slides, Corning®
Microscope slides, Corning®
Plain slides, Colourless, Glass, 75×38 mm
Catalog # 734-4036
Supplier:   Corning
Microscope slides, Corning®
Catalog # 734-4036
Supplier:  Corning
Microscope slides, Corning®
Microscope slides, Corning®
Plain slides, Colourless, Glass, 75×38 mm
Price
317,00 €
Log in for availability
1 * 720 items
Product Details & Documents
These slides are either frosted on one side, on both sides, or plain.
  • Clarity reinforced water-white glass
  • Frosted slides are sandblasted, easy to write on and highly legible
  • Each slide is inspected for chips and rough edge before being shipped
Specifications
  • Colour:
    Colourless
  • Description:
    Plain slides
  • L×W:
    75×38 mm
  • Material:
    Glass
  • Seq:
    04