Orders LinkContactUsLinkComponent
Microscope slides, Corning®
Plain slides, Colourless, Glass, 75×25 mm
  734-4035
 :   Corning
Microscope slides, Corning®
  734-4035
 :  Corning
Microscope slides, Corning®
Microscope slides, Corning®
Plain slides, Colourless, Glass, 75×25 mm
386,00 €
1 * 1.440 items
These slides are either frosted on one side, on both sides, or plain.
  • Clarity reinforced water-white glass
  • Frosted slides are sandblasted, easy to write on and highly legible
  • Each slide is inspected for chips and rough edge before being shipped
  • Colour:
    Colourless
  • Description:
    Plain slides
  • L×W:
    75×25 mm
  • Material:
    Glass
  • Seq:
    01