Filter sets
Filter set 02, excitation G 365, beam splitter FT 395, emission LP 420
ZEIS4880029901000
: ZEISS
- Description:Filter set 02, excitation G 365, beam splitter FT 395, emission LP 420
- Seq:01