Orders LinkContactUsLinkComponent
Screw caps for internal thread sample tubes
Screw caps with silicone washer, Green
  SIMPT500GOS
Screw caps for internal thread sample tubes
  SIMPT500GOS
Screw caps for internal thread sample tubes
Screw caps for internal thread sample tubes
Screw caps with silicone washer, Green
246,00 €
1 * 1.000 items
These polypropylene screw caps are to be used with T500 series internal thread sample tubes.
  • Made of polypropylene
  • Cap with silicone washer for leakproof seal
  • Long skirt and super-fast thread design for easy seal
  • Autoclavable
Caps are available with a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures. The cap features a long skirt and a super-fast thread design allowing it to be removed or screwed on with a single turn.
These silicone washer caps will guarantee a positive leakproof seal at all temperatures.
 : Tubes T500 series sold separately.
  • Colour:
    Green
  • Description:
    Screw caps with silicone washer