6700 Split beam, visiblespectrophotometer, 320-1100nm Model 6700 visible range, 4nm SBW spectrophotometer fitted with single 10 x 10 mm cuvette holder, 1GB of internal memory, 1GB SD/USB memory card, 100 disposable cuvettes, instruction manual, power 1 * 1 items
Catalog # 634-0583
Supplier: Cole-Parmer
Frequently Bought Together