Pipette tips
Micro, ultrafine, 0,1 - 10 µl, +, 32 mm, Natural, 96 tips/rack; 12 racks/case
Catalog # ALPCLW1136
Supplier: Alpha Laboratories
Jump to:
Product Details & Documents
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
- Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
- Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
- With graduations to visually check volume
- Suitable for all major pipettor brands
Specifications
- Colour code:Natural
- Packed:96 tips/rack; 12 racks/case
- Sterile:+
- Volume:0,1 - 10 µl
- Length:32 mm
- Tip style:Micro, ultrafine