Caps, LABOCAP, without grip
28-30 mm, Red
: LUDI
Made of anodised aluminum with stainless steel spring clip.
- Springs clamp against the outer wall of the vessel
- Anodised and coloured
- Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.
- For Ø ext.:28-30 mm
- Colour:Red
- Seq:24