Orders LinkContactUsLinkComponent
Objectives
N-Planachromatic, M27, N.A. of 1,25, WD of 0,29 mm
  ZEIS4209909900000
 :   ZEISS
Objectives
  ZEIS4209909900000
 :  ZEISS
Objectives
Objectives
N-Planachromatic, M27, N.A. of 1,25, WD of 0,29 mm
1 * 1 items
  • Magnification:
    100×
  • Description:
    N-Planachromatic, M27, N.A. of 1,25, WD of 0,29 mm
  • Seq:
    02