Orders LinkContactUsLinkComponent
Objectives
Objectives
N-Planachromatic, M27, Ph3, N.A. of 1,25, WD of 0,29 mm
Catalog # ZEIS4209919910000
Supplier:   ZEISS
Objectives
Catalog # ZEIS4209919910000
Supplier:  ZEISS
Objectives
Objectives
N-Planachromatic, M27, Ph3, N.A. of 1,25, WD of 0,29 mm
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Magnification:
    100×
  • Description:
    N-Planachromatic, M27, Ph3, N.A. of 1,25, WD of 0,29 mm
  • Seq:
    03