Digital Solutions
Applications
Support
About
Slovenia
ContactUsLinkComponent
Phosphate Standard FS 6X3ml 1 * 18 ml
Catalog # DIAS152109910030
Supplier:
DIAGNOSTIC SYSTEMS DIASYS
Certificates
Expand image
Phosphate Standard FS 6X3ml 1 * 18 ml
Catalog # DIAS152109910030
Supplier:
DIAGNOSTIC SYSTEMS DIASYS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals