Kocuria rhizophila 2.5 ml Growth Promotion Suspensions, USP/EP compliant, provide a QC challenge to each lot of medium whether purc hased commerically or prepared in-house. Population level of <100 Colony Forming Units (CFUs) per each 0.1 ml dose. 1 * 2,5 ml
NAMSGP25-05
: CROSSTEX
Frequently Bought Together