Permanent marker
Permanent marker, fiber tip, Red
Catalog # KOEH2-5054
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Pens, permanent marking, red, 1 * 6 items Catalog #811-0057
Product 811-0057 is similar to KOEH2-5054 but may have a different manufacturer and/or packaging.
Product Details & Documents
This versatile paint marker dispense a fast-drying, high-gloss enamel paint for permanent marking, lettering, coding and numbering of both interior and exterior surfaces of metal, plastic, printed circuit boards, glass, wood, paper, leather, textiles, etc. This marker can write on porous or non-porous; rough or smooth; wet, oily or dry surfaces.
- Marks are permanent
- Withstand heat and weathering under adverse conditions
- Available in different lead-free colours
- With long-lasting replaceable fiber tips
Specifications
- Colour:Red
- Description:Permanent marker, fiber tip
- Seq:01