Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

2-Chlorocyclohexanone ≥97% stabilised

Glass bottle for solids, 50 g
Catalog # A18613.18
Irritant
2-Chlorocyclohexanone ≥97% stabilised
Catalog # A18613.18
CAS Number:  822-87-7
2-Chlorocyclohexanone ≥97% stabilised
2-Chlorocyclohexanone ≥97% stabilised

Glass bottle for solids, 50 g

CAS Number:  822-87-7
Price
189,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Stabilized with calcium carbonate/magnesium oxide

Purity: 97% CAS No.: 822-87-7 Molecular Formula: C6H9ClO Formula Weight: 132.59 Storage Temperature: -30°C to -10°C Flash Point: 82°(180°F) Refractive Index: 1.4840 Melting Point: 22-24° Boiling Point: 82-83°/10mm Beilstein Reference: 774100 MDL No.: MFCD00001626

  • CAS Number:822-87-7
  • Formula:C₆H₉ClO
  • MDL Number:MFCD00001626
  • Molecular Weight:132.59 g/mol
  • Boiling Pt:82…83 °C (10 mmHg)
  • Melting Pt:22…24 °C
  • Density:1.161
  • Flash Pt:82 °C (180 °F)
  • Storage Temperature:Freezer
  • EINECS:212-505-5
  • ADR:9,
  •  :Stabilised with calcium carbonate/magnesium oxide
  • UN:3335
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.