Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Chlorocyclohexanone ≥97% stabilised

2-Chlorocyclohexanone ≥97% stabilised

Glass bottle for solids, 10 g
Catalog # A18613.09
Irritant
2-Chlorocyclohexanone ≥97% stabilised
Catalog # A18613.09
CAS Number:  822-87-7
2-Chlorocyclohexanone ≥97% stabilised
2-Chlorocyclohexanone ≥97% stabilised

Glass bottle for solids, 10 g

CAS Number:  822-87-7
Price
58,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Stabilized with calcium carbonate/magnesium oxide

Purity: 97% CAS No.: 822-87-7 Molecular Formula: C6H9ClO Formula Weight: 132.59 Storage Temperature: -30°C to -10°C Flash Point: 82°(180°F) Refractive Index: 1.4840 Melting Point: 22-24° Boiling Point: 82-83°/10mm Beilstein Reference: 774100 MDL No.: MFCD00001626

  • CAS Number:822-87-7
  • Formula:C₆H₉ClO
  • MDL Number:MFCD00001626
  • Molecular Weight:132.59 g/mol
  • Boiling Pt:82…83 °C (10 mmHg)
  • Melting Pt:22…24 °C
  • Density:1.161
  • Flash Pt:82 °C (180 °F)
  • Storage Temperature:Freezer
  • EINECS:212-505-5
  • ADR:9,
  •  :Stabilised with calcium carbonate/magnesium oxide
  • UN:3335
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications