Electronic dispenser Ripette® genX for high-precision dosage-applications, select and dispense by digital display volume range from 1µL to 50000 µL, with Euro AC-adapter 1 * 1 items
Catalog # RITT42100-0000
Supplier: Ritter, part of Avantor
Frequently Bought Together