Orders LinkContactUsLinkComponent
Pycnometer with thermometer, adjusted
Pycnometer with thermometer, adjusted
10 ml, 10/19
Catalog # HECH42572016
Pycnometer with thermometer, adjusted
Catalog # HECH42572016
Pycnometer with thermometer, adjusted
Pycnometer with thermometer, adjusted
10 ml, 10/19
Price
151,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
  • With ground-in capillary stopper
  • Very good chemical resistance
  • Very good temperature resistance
It includes pycnometer bottle and ground-in thermometer joint size 10/19 with blue filling.
Certifications: DIN ISO 3507
Specifications
  • NS:
    10/19
  • Volume (nom.):
    10 ml