Powder samplers, PharmaPicker
PharmaPicker 1, Stainless steel V4A, 600 mm, 0 – 1,25 ml
Catalog # BURK5346-1000
Supplier: BURKLE
Jump to:
Product Details & Documents
PharmaPicker is designed for sampling very small quantities of highly potent or active powders for analysis. The smallest sample quantities can be collected with the PharmaPicker.
- Available separately or in a handy transport box
- Electro-polished stainless steel
- Can be extended repeatedly
Delivery information: PharmaPicker case includes collection cylinders in both sizes, extension rods of 50 and 100 cm, a handle, volume tips (alternative sample volumes on request), and a transport box.
Specifications
- Volume:0 – 1,25 ml
- Description:PharmaPicker 1
- Length:600 mm
- Material:Stainless steel V4A
Accessories
Product Information
- Catalog #AvailabilityPrice
- Volume heads for PharmaPicker, capacity 0,60 mlVolume heads for PharmaPicker, capacity 0,60 mlBURK5346-1060Log in for availability1 * 1 items - 92,40 €
Product Information
- Catalog #AvailabilityPrice
- Collection cylinder for container volume 0,10 - 1,25 mlCollection cylinder for container volume 0,10 - 1,25 mlBURK5346-1001Log in for availability1 * 1 items - 303,00 €
Product Information
- Catalog #AvailabilityPrice
- Volume heads for PharmaPicker, capacity 0,10 mlVolume heads for PharmaPicker, capacity 0,10 mlBURK5346-1010Log in for availability1 * 1 items - 92,40 €
Product Information
- Catalog #AvailabilityPrice
- Collection cylinder for container volume 1,25 - 2,50 mlCollection cylinder for container volume 1,25 - 2,50 mlBURK5346-2001Log in for availability1 * 1 items - 289,00 €
Product Information
- Catalog #AvailabilityPrice
- Extension rod PharmaPicker, 1000 mm
Product Information
- Catalog #AvailabilityPrice
- Extension rod PharmaPicker, 500 mm
Product Information
- Catalog #AvailabilityPrice
- Volume heads for PharmaPicker, capacity 0,20 mlVolume heads for PharmaPicker, capacity 0,20 mlBURK5346-1020Log in for availability1 * 1 items - 92,40 €
Product Information
- Catalog #AvailabilityPrice
- PharmaPicker handle
Product Information
- Catalog #AvailabilityPrice
- Volume heads for PharmaPicker, capacity 0,80 mlVolume heads for PharmaPicker, capacity 0,80 mlBURK5346-1080Log in for availability1 * 1 items - 92,40 €
Product Information
- Catalog #AvailabilityPrice
- Volume heads for PharmaPicker, capacity 2,50 mlVolume heads for PharmaPicker, capacity 2,50 mlBURK5346-2250Log in for availability1 * 1 items - 92,40 €
Product Information
- Catalog #AvailabilityPrice
- Volume heads for PharmaPicker, 1 ml
Product Information
- Catalog #AvailabilityPrice
- Volume heads for PharmaPicker, 2 ml